Learn More
Fisherbrand™ Plastic PP Two-Part Syringes, Luer Lock
HPLC certified, non-sterile, two-part disposable syringes made of solvent robust polypropylene.
Brand: Fisherbrand™ 02 36 2399
Description
- Plastic Disposable Syringe
- Polypropylene
- Luer Lock
- Two-part, all-plastic construction eliminates the need for rubber or synthetic plunger gaskets
- No silicone or oil lubricant is required in the barrel
Specifications
Polypropylene | |
2 mL | |
Luer Lock |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Fisherbrand™ Plastic PP Two-Part Syringes, Luer Lock > Volume: 2mL
Spot an opportunity for improvement?Share a Content Correction