Learn More
Sartorius™ 11302 Series Non-Sterile Cellulose Nitrate (CN) Membrane Filters
11302 Series Non-Sterile Cellulose Nitrate (CN) Membrane Filters are hydrophilic, have high flow rates thanks to their symmetrical structure, and are compatible with aqueous solutions (pH 4 – 8), hydrocarbons, and several other organic solvents.
Brand: Sartorius™ 11302--13------N
Code : NEW
Additional Details : Weight : 0.10000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Description
- High, non-specific absorption
- Hydrophilic, with high flow rates
- Available in a range of pore sizes and diameters
- Ideal for several laboratory applications including particle retention, cell capture, cell retention, and cell filtration
Specifications
Autoclavable | |
White | |
Membrane Filter | |
Cellulose Nitrate | |
3 μm | |
430 mL/min./cm2/bar | |
100 Pk. | |
5 mil | |
Membrane Filter | |
13 mm |
0.5 bar | |
Single-use | |
Disc | |
Cellulose Nitrate | |
Non-sterile | |
For many general laboratory applications where a membrane with a high non-specific adsorption is suitable | |
100 Pk. | |
130 μm | |
0.5 in. | |
13 mm |
Safety and Handling
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P230-Keep wetted with .
- P233-Keep container tightly closed.
- P234-Keep only in original container.
- P243-Take precautionary measures against static discharge.
- P370+P378-In case of fire: Use to extinguish.
- P372-Explosion risk in case of fire.
- P375-Fight fire remotely due to the risk of explosion.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.