missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 11mm Glass Snap Top Vials - Routine Range

Use routine snap vials for your optimum high throughput chromatographic analysis.

Brand:  Thermo Scientific™ 60180A-RV11-1P

 View more versions of this product

Product Code. 17199548

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Use the Thermo Scientific™ 11mm Glass Snap Top Vials for optimum performance for standard applications and high throughput analyses. These autosampler vials exhibit easy handling and high solvent compatibility for the mainstream HPLC and GC applications. Recommended workflows / applications include:

  • Chromatographic applications aligning with all pharmacopeias
  • Lower risk of contamination, degradation and adsorption
  • Standard concentrations
  • Perfect autosampler fit
  • High throughput analysis

Specifications

Specifications

2mL vial, clear glass, 11mm snap top, wide opening, ID patch, 100/Pack
Glass
Snap Cap
Yes
Clear
2 mL
11 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers