Learn More
Watson-Marlow Pumps 120 Series Drive with Pumphead
100 Series Pumps are small and stackable, saving valuable space in cleanroom, LAF and biosafety cabinet installations, capable of flow rates up to 190 mL/min.
Brand: Watson-Marlow Pumps
Additional Details : Weight : 1.90000kg
Description
101 series pumps are small and stackable lab pumps for handling valuable and delicate fluids without contamination. Save valuable space in cleanrooms, LAF and biosafety cabinets.
- The 100 series is an ideal pump for maximizing yield in fermentation and cell culture applications.
- Ultra-compact and stackable for multiple feeds such as pH control, antifoam, nutrient and buffer addition, the 120 offers the ideal combination of size and technology to optimize your process.
- The 100 series pump is suited to single-use systems.
- Error-free tube change takes only seconds.
- Biopharmaceutical processes require accurate and repeatable pumping that is easy to validate.
- With a 2,000:1 speed range, the 100 series pump delivers the flexibility and purity so vital to drug trials.
- A keypad lock secures your pumping parameters for consistent and repeatable performance.
- 120R single channel, two roller pumphead for continuous tubing
- 114DV flip-top single-channel pumphead for continuous tubing
- 400D1 single-channel pumphead for continuous tubing
- 400DM2 twin-channel pumphead for manifold tubing
- 400DM3 three-channel pumphead for manifold tubing
Specifications
Analog | |
IP31 | |
30 psi | |
120F | |
90/264 V |
LED | |
0.002 to 190 mL/min. | |
220 rpm | |
114DV |