missing translation for 'onlineSavingsMsg'
Learn More
Learn More
124 mV Redox Oxidation/Reduction (ORP) Standard at 25°C, Reagecon
Acts as a control or reference material, rather than as a calibration standard because redox oxidation/reduction (ORP) is an absolute measurement and the sensing electrode does not require calibration.
Brand: Reagecon RS124
This item is not returnable.
View return policy
Description
- Redox Reference Standard, Redox Reference Material.
- Acts as a control or reference material rather than a calibration standard because the redox reaction is an absolute measurement and the sensing electrode does not require calibration.
- Controls the functionality of the sensing and reference electrode, the analysts technique, environmental conditions and the operation of the measuring meter.
- This 124 mV Redox (ORP) Standard is accurate to ± 5mV, has guaranteed stability throughout its entire shelf life, even after opening the bottle.
- The Value of 124 mV quoted refers to the potentials of the platinum electrode v Ag/AgCl reference electrode (3M KCl).
Specifications
Accuracy ± 5mV at 25°C | |
HDPE Bottle GL45 | |
124 mV (± 5mV) |
124 mV (± 5mV) | |
500 mL | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
RS12424J1 | Certificate of Analysis | 26/11/2024 | RS124 |
RS12423J1 | Certificate of Analysis | 20/08/2024 | RS124 |
RS12423J1 (2) | Certificate of Analysis | 15/03/2024 | RS124 |
RS12423J1 (1) | Certificate of Analysis | 15/03/2024 | RS124 |
RS12422G1 | Certificate of Analysis | 05/09/2023 | RS124 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
124 mV Redox Oxidation/Reduction (ORP) Standard at 25°C, Reagecon > 500 mL
Spot an opportunity for improvement?Share a Content Correction