missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 13mm PP Screw Cap, 13-425 thread

Polypropylene caps are chemically inert and suitable for most chromatography applications.

Brand:  Fisherbrand™ 13 08 0639

 View more versions of this product

Product Code. 11717607

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Description

Description

  • 13mm
  • PP Screw Cap
  • Center hole
TRUSTED_SUSTAINABILITY
Specifications

Specifications

White
Polypropylene
Center Hole Screw Cap
13 mm
100
ND13
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers