Learn More
Corning™ 1536-Well Polystyrene NBS™-Treated Microplates
Proprietary nonbinding surface is ideal for homogeneous assays with sticky enzymes or for reducing nonspecific binding to microplate wells
Brand:  Corning™ 3729BC
 View more versions of this product
Product Code. 10052443
  • £1778.82 / Pack of 50
Estimated Shipment: 01-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
  • Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
  • Recommended working volume up to 8 μL
  • Round well bottom for reduced air entrapment and improved CV values and Z factor
  • Raised well bottom for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.
Order Info
Packaged in bags of ten
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
32 x 48
Polystyrene
8 μL
Non-binding
Flat Bottom (F-well)
50/Cs.
White
Without Lid
White
1536
Non-sterile
12.5 μL
Generic barcodes
10/Pk.