missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 1536-Well Polystyrene NBS™-Treated Microplates
Proprietary nonbinding surface is ideal for homogeneous assays with sticky enzymes or for reducing nonspecific binding to microplate wells
£1232.63 - £1723.37
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 32 x 48 |
No. of Wells | 1536 |
Volume (Metric) Working | 8 μL |
Product Code | Brand | Color | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Includes | Price | Quantity & Availability | ||||
10143613
|
Corning™
3728BC |
Black | Generic barcodes |
£1279.30
Pack of 50 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
17720708
|
Corning™
4572 |
White, Clear Bottom | Barcode |
£1510.82
Pack of 50 |
In Stock
Log in to see stock. |
||||
10052443
|
Corning™
3729BC |
White | Generic barcodes |
£1723.37
Pack of 50 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||||
17790698
|
Corning™
4567 |
Black, Clear Bottom | Barcode |
£1510.82
Pack of 50 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||||
10189472
|
Corning™
3895 |
Black, Clear Bottom | Without barcode |
£1658.70
Pack of 50 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||||
10027322
|
Corning™
3728 |
Black | Without barcode |
£1232.63
Pack of 50 |
In Stock
Log in to see stock. |
||||
Description
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
- Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
- Recommended working volume up to 8 μL
- Round well bottom for reduced air entrapment and improved CV values and Z factor
- Raised well bottom for higher sensitivity
- Flood reservoir on four sides to reduce instrument contamination
- Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.
Order Info
Packaged in bags of ten
Specifications
Standard | |
32 x 48 | |
8 μL | |
12.5 μL | |
50/Cs. |
Without Lid | |
1536 | |
Non-sterile | |
Flat Bottom (F-well) | |
10/Pk. |