missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 E-Poli Crucible Tongs with Plastic Handle PVC

Grips well

Brand:  Bochem™ 2162

 View more versions of this product

Product Code. 10336241

  • £16.45 / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With plastic handle PVC
  • Maxium temperature 145°C
Specifications

Specifications

250 mm
18/10 E-Poli
Crucible Tongs
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bochem™ 18/10 E-Poli Crucible Tongs with Plastic Handle PVC > Length/Diameter: 250/5mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.