missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Dissecting Needle

For use with Bochem needle holder Kolle 12000. Bochem™ 18/10 Stainless Steel Dissecting Needle is manufactured from stainless steel.

Brand:  Bochem™ 12003

 View more versions of this product

Product Code. 11865101

  • £15.30 / Pack of 10
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Type 1: eye
  • Type 2: needle
  • Type 3: lancet
  • For use with Bochem needle holder Kolle 12000
  • Manufactured from stainless steel
Specifications

Specifications

4 mm
Stainless Steel (18/10)
Type 1 Eye Holder KOLLE 12000
100 mm
Eye
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers