missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Double Spatulas, Round/Bent Ends, Type 1

Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Double Spatulas serve many uses in the lab.

Brand:  Bochem™ 3181

Code : A0

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 10495611

  • £4.45 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Double spatula
  • Manufactured from 18/10 stainless steel
Specifications

Specifications

1 Ea.
Double Ended Spatula
5 mm
Stainless Steel (18/10)
Round/Bent Ends, Type 1
10 mm
5 mm
185 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers