missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Micro Double Spatulas

Bent, double-ended, micro spatulas constructed of stainless steel. Bochem™ Micro Double Spatulas, Bent are slim, chemically resistant, and well-suited for transferring small material amounts.

Brand:  Bochem™ 3021G

 View more versions of this product

Product Code. 10373611

  • £3.65 / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Durable 18/10 stainless steel construction.
  • Double-ended for versatility.
  • Slim ends for easy container access.
  • One rounded end and one flat end.
Specifications

Specifications

1 Ea.
Double Ended Micro Spatula
5 mm
2mm
Stainless Steel (18/10)
Bent
40 mm
Stainless Steel (18/10)
130 x 5 mm
130 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers