missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Poly Spoons

Manufactured from 18/10 stainless steel

Brand:  Bochem™ 3412

 View more versions of this product

Product Code. 10791601

  • £18.25 / Each
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Spoon comes in three different types
  • Autoclavable
  • Made of 18/10 stainless steel
  • Multi-purpose spoon for a wide range of uses
Specifications

Specifications

1 Ea.
Poly Spoon
35 mm
Stainless Steel (18/10)
300 x 20 mm
300 mm
Autoclavable
Multi-purpose spoon for a wide range of uses
20 mm
8mm
Stainless Steel (18/10)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers