Learn More
1H,1H,2H,2H-Perfluorooctyltrichlorosilane, 97%, Thermo Scientific Chemicals
£76.40 - £295.00
Chemical Identifiers
CAS | 78560-45-9 |
---|---|
Molecular Formula | C8H4Cl3F13Si |
Molecular Weight (g/mol) | 481.534 |
MDL Number | MFCD00042363 |
InChI Key | PISDRBMXQBSCIP-UHFFFAOYSA-N |
Synonym | trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, 1h,1h,2h,2h-perfluorooctyltrichlorosilane, silane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, tridecafluoro-1,1,2,2-tetrahydrooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluorooctyl silane, 2-tridecafluorohexyl ethyltrichlorosilane, 1h,1h,2h,2h-perfluorooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluoro-n-octyl silane, trichloro 1h,1h,2h,2h-tridecafluoro-n-octyl silane |
PubChem CID | 123578 |
IUPAC Name | trichloro(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane |
SMILES | C(C[Si](Cl)(Cl)Cl)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11329285
|
Thermo Scientific Alfa Aesar
L16606.09 |
10 g |
£76.40
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11339285
|
Thermo Scientific Alfa Aesar
L16606.18 |
50 g |
£295.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1H,1H,2H,2H-Perfluorooctyltrichlorosilane is used in the silanization of silicon wafer. Self assembled monolayers treated with this, acts as an anti adhesive layer and release of cured poly(dimethyl siloxanes). It is also used to fabricate chemical surface patterns of self assembled monolayers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1H,1H,2H,2H-Perfluorooctyltrichlorosilane is used in the silanization of silicon wafer. Self assembled monolayers treated with this, acts as an anti adhesive layer and release of cured poly(dimethyl siloxanes). It is also used to fabricate chemical surface patterns of self assembled monolayers.
Solubility
Miscible with tetrahydrofuran, tetrhydropyran, toluene and other organic solvents.
Notes
Incompatible with water, strong oxidizing agents, alcohols, acids and bases.
Chemical Identifiers
78560-45-9 | |
481.534 | |
PISDRBMXQBSCIP-UHFFFAOYSA-N | |
123578 | |
C(C[Si](Cl)(Cl)Cl)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
C8H4Cl3F13Si | |
MFCD00042363 | |
trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, 1h,1h,2h,2h-perfluorooctyltrichlorosilane, silane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, tridecafluoro-1,1,2,2-tetrahydrooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluorooctyl silane, 2-tridecafluorohexyl ethyltrichlorosilane, 1h,1h,2h,2h-perfluorooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluoro-n-octyl silane, trichloro 1h,1h,2h,2h-tridecafluoro-n-octyl silane | |
trichloro(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane |
Specifications
192°C | |
C8H4Cl3F13Si | |
UN2987 | |
87°C (189°F) | |
Miscible with tetrahydrofuran,tetrhydropyran,toluene and other organic solvents. | |
C(C[Si](Cl)(Cl)Cl)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F | |
481.534 | |
481.55 | |
1.352 | |
1.638 |
78560-45-9 | |
MFCD00042363 | |
6154526 | |
trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, 1h,1h,2h,2h-perfluorooctyltrichlorosilane, silane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, tridecafluoro-1,1,2,2-tetrahydrooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluorooctyl silane, 2-tridecafluorohexyl ethyltrichlorosilane, 1h,1h,2h,2h-perfluorooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluoro-n-octyl silane, trichloro 1h,1h,2h,2h-tridecafluoro-n-octyl silane | |
PISDRBMXQBSCIP-UHFFFAOYSA-N | |
trichloro(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane | |
123578 | |
97% | |
Moisture sensitive | |
1H,1H,2H,2H-Perfluorooctyltrichlorosilane |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 278-947-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only