missing translation for 'onlineSavingsMsg'
Learn More
1H-Indole-2-methanol, 96%, Thermo Scientific Chemicals Product Code.: 15582373

1H-Indole-2-methanol, 96%, Thermo Scientific Chemicals

Product Code. 15582373
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15582373

Brand: Thermo Scientific Acros 459810010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24621-70-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.17
InChI Key XEEANGGQJOWRTG-UHFFFAOYSA-N
Synonym 1h-indol-2-yl methanol, 1h-indole-2-methanol, 1h-indol-2-yl-methanol, 2-hydroxymethylindole, 2-hydroxymethyl indole, hydroxymethylindol, 2-indolemethanol, indole-2-methanol, indol-2-ylmethan-1-ol
PubChem CID 98783
IUPAC Name 1H-indol-2-ylmethanol
SMILES C1=CC=C2C(=C1)C=C(N2)CO

Specifications

Melting Point 72.0°C to 78.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 147.17
Percent Purity 96%
Chemical Name or Material 1H-Indole-2-methanol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0419148 Certificate TypeCertificate of Analysis Date14/11/2022 Catalog Number
compliance-icons
Product Identifier
  • 1H-Indole-2-methanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1H-Indole-2-methanol, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.