Learn More
IBA Lifesciences Strep-Tactin™ Sepharose™ Column
Strep-Tactin™ Sepharose™ column is a Gravity flow column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
Brand:  IBA Lifesciences 2-1202-001
 View more versions of this product
Product Code. 12886436
  • £76.50 / 1mL
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Strep-Tactin™ Sepharose™ columns contains 4% agarose coupled with the streptavidin variant Strep-Tactin™ and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
  • Due to the low concentrated agarose, it is particularly suitable for large proteins, especially if the affinity to Strep-Tactin™XT 4Flow™ is too high.
  • The wide-meshed polymer allows for the simple attachment of larger proteins to the stationary phase.
  • To reduce the waiting period during sample and buffer application, the use of the application aid WET FRED is recommended. 
Specifications
Specifications
1 mL