missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,4S)-(-)-2-BOC-2,5-diazabicyclo[2.2.1]heptane, 95%, Thermo Scientific Chemicals
£143.00 - £620.00
Chemical Identifiers
CAS | 113451-59-5 |
---|---|
Molecular Formula | C10H18N2O2 |
Molecular Weight (g/mol) | 198.26 |
MDL Number | MFCD01569250 |
InChI Key | UXAWXZDXVOYLII-YUMQZZPRSA-N |
Synonym | 1s,4s-2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-tert-butyl 2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-n-tert-butoxycarbonyl-2,5-diazabicyclo 2.2.1 heptane, 1s,4s---2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptan-2-carboxylate, 1s,4s-2,5-diaza-bicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, 1s,4s-n-boc-2,5-diazabicyclo 2.2.1 heptane, 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, tert-butyl 1s,4s---2,5-diazabicyclo 2.2.1 heptane-2-carboxylate |
PubChem CID | 11521263 |
IUPAC Name | tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CC2CC1CN2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10693975
|
Thermo Scientific Acros
398040010 |
1 g | Glass bottle |
£143.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10597054
|
Thermo Scientific Acros
398040050 |
5 g | Glass bottle |
£620.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
113451-59-5 | |
198.26 | |
UXAWXZDXVOYLII-YUMQZZPRSA-N | |
11521263 | |
CC(C)(C)OC(=O)N1CC2CC1CN2 |
C10H18N2O2 | |
MFCD01569250 | |
1s,4s-2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-tert-butyl 2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-n-tert-butoxycarbonyl-2,5-diazabicyclo 2.2.1 heptane, 1s,4s---2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptan-2-carboxylate, 1s,4s-2,5-diaza-bicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, 1s,4s-n-boc-2,5-diazabicyclo 2.2.1 heptane, 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, tert-butyl 1s,4s---2,5-diazabicyclo 2.2.1 heptane-2-carboxylate | |
tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate |
Specifications
113451-59-5 | |
100.0 | |
White to Beige | |
94% min. (GC) | |
MFCD01569250 | |
1s,4s-2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-tert-butyl 2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-n-tert-butoxycarbonyl-2,5-diazabicyclo 2.2.1 heptane, 1s,4s---2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptan-2-carboxylate, 1s,4s-2,5-diaza-bicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, 1s,4s-n-boc-2,5-diazabicyclo 2.2.1 heptane, 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, tert-butyl 1s,4s---2,5-diazabicyclo 2.2.1 heptane-2-carboxylate | |
UXAWXZDXVOYLII-YUMQZZPRSA-N | |
tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate | |
11521263 | |
95% | |
(1S,4S)-(-)-2-BOC-2,5-diazabicyclo[2.2.1]heptane |
94.0 | |
74.0°C to 76.0°C | |
Authentic | |
C10H18N2O2 | |
-41° to -47° (20°C, 589nm) (C=1, chloroform) | |
Solubility in water: insoluble | |
CC(C)(C)OC(=O)N1CC2CC1CN2 | |
198.26 | |
198.26 | |
Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- (1S,4S)-(-)-2-BOC-2,5-diazabicyclo¢2.2.1!heptane
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only