Learn More
2-(1-Adamantyl)acetic acid, 97%, Thermo Scientific™
£8.15 - £52.95
Chemical Identifiers
CAS | 4942-47-6 |
---|---|
Molecular Formula | C12H17O2 |
Molecular Weight (g/mol) | 193.27 |
MDL Number | MFCD00074728 |
InChI Key | AOTQGWFNFTVXNQ-UHFFFAOYSA-M |
Synonym | 1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane |
PubChem CID | 123221 |
SMILES | [O-]C(=O)CC12CC3CC(CC(C3)C1)C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10567074
|
Thermo Scientific Maybridge
BTB13005DA |
1 g |
£8.15
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10442624
|
Thermo Scientific Maybridge
BTB13005EA |
10 g |
£33.40
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10632465
|
Thermo Scientific Maybridge
BTB13005EB |
25 g |
£52.95
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
4942-47-6 | |
193.27 | |
AOTQGWFNFTVXNQ-UHFFFAOYSA-M | |
123221 |
C12H17O2 | |
MFCD00074728 | |
1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane | |
[O-]C(=O)CC12CC3CC(CC(C3)C1)C2 |
Specifications
4942-47-6 | |
100.0 | |
Amber Glass Bottle | |
MFCD00074728 | |
AOTQGWFNFTVXNQ-UHFFFAOYSA-M | |
2-(adamantan-1-yl)acetate | |
123221 | |
97% |
96.5 | |
97% | |
C12H17O2 | |
1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane | |
[O-]C(=O)CC12CC3CC(CC(C3)C1)C2 | |
193.27 | |
194.27 | |
2-(1-Adamantyl)acetic acid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning