Learn More
2,2,2-Trimethylthioacetamide, 97%, Thermo Scientific Chemicals
£42.10 - £249.00
Chemical Identifiers
CAS | 630-22-8 |
---|---|
Molecular Formula | C5H11NS |
Molecular Weight (g/mol) | 117.21 |
MDL Number | MFCD09742834 |
InChI Key | FJZJUSOFGBXHCV-UHFFFAOYSA-N |
Synonym | 2,2,2-trimethylthioacetamide, 2,2-dimethylthiopropionamide, neopentanethioamide, propanethioamide, 2,2-dimethyl, thiopivalamide, 2,2-dimethyl-thiopropionamide, 2,2-dimethylthio propionamide, propanethioamide,2,2-dimethyl, 2-methyl-2-methylthiopropionamide, propionamide, 2-methyl-2-methylthio |
PubChem CID | 3031130 |
IUPAC Name | 2,2-dimethylpropanethioamide |
SMILES | CC(C)(C)C(=S)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11370036
|
Thermo Scientific Alfa Aesar
H32092.03 |
1 g |
£42.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11360036
|
Thermo Scientific Alfa Aesar
H32092.09 |
10 g |
£249.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,2,2-Trimethylthioacetamide, is used as a pharmaceutical and organic intermediate, an important amino protective agent, pharmaceutical and synthetic organic make intermediates, organic synthesis used to introduce t- Boc protect gene species.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2,2-Trimethylthioacetamide, is used as a pharmaceutical and organic intermediate, an important amino protective agent, pharmaceutical and synthetic organic make intermediates, organic synthesis used to introduce t- Boc protect gene species.
Solubility
Soluble in water (10 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
630-22-8 | |
117.21 | |
FJZJUSOFGBXHCV-UHFFFAOYSA-N | |
3031130 | |
CC(C)(C)C(=S)N |
C5H11NS | |
MFCD09742834 | |
2,2,2-trimethylthioacetamide, 2,2-dimethylthiopropionamide, neopentanethioamide, propanethioamide, 2,2-dimethyl, thiopivalamide, 2,2-dimethyl-thiopropionamide, 2,2-dimethylthio propionamide, propanethioamide,2,2-dimethyl, 2-methyl-2-methylthiopropionamide, propionamide, 2-methyl-2-methylthio | |
2,2-dimethylpropanethioamide |
Specifications
630-22-8 | |
C5H11NS | |
2,2,2-trimethylthioacetamide, 2,2-dimethylthiopropionamide, neopentanethioamide, propanethioamide, 2,2-dimethyl, thiopivalamide, 2,2-dimethyl-thiopropionamide, 2,2-dimethylthio propionamide, propanethioamide,2,2-dimethyl, 2-methyl-2-methylthiopropionamide, propionamide, 2-methyl-2-methylthio | |
FJZJUSOFGBXHCV-UHFFFAOYSA-N | |
2,2-dimethylpropanethioamide | |
3031130 | |
97% |
113°C to 117°C | |
MFCD09742834 | |
Soluble in water (10g/L) (25°C). | |
CC(C)(C)C(=S)N | |
117.21 | |
117.21 | |
2,2,2-Trimethylthioacetamide |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only