Learn More
2-(2-Aminoethoxy)ethanol, 98%, Thermo Scientific Chemicals
£24.50 - £312.00
Chemical Identifiers
CAS | 929-06-6 |
---|---|
Molecular Formula | C4H11NO2 |
Molecular Weight (g/mol) | 105.14 |
InChI Key | GIAFURWZWWWBQT-UHFFFAOYSA-N |
Synonym | 2-2-aminoethoxy ethanol, diglycolamine, 2-aminoethoxyethanol, ethanol, 2-2-aminoethoxy, diethylene glycol amine, 2-2-hydroxyethoxy ethylamine, diethylene glycol monoamine, 2-amino-2'-hydroxydiethyl ether, 1-amino-2-2-hydroxyethoxy ethane, 5-hydroxy-3-oxapentylamine |
PubChem CID | 13578 |
IUPAC Name | 2-(2-aminoethoxy)ethanol |
SMILES | C(COCCO)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10264700
|
Thermo Scientific Acros
103631000 |
100 mL | Glass bottle |
£24.50
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10519630
|
Thermo Scientific Acros
103630010 |
1 L | Glass bottle |
£130.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11965271
|
Thermo Scientific Acros
103630025 |
2.5 L | Glass bottle |
£312.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
929-06-6 | |
105.14 | |
2-2-aminoethoxy ethanol, diglycolamine, 2-aminoethoxyethanol, ethanol, 2-2-aminoethoxy, diethylene glycol amine, 2-2-hydroxyethoxy ethylamine, diethylene glycol monoamine, 2-amino-2'-hydroxydiethyl ether, 1-amino-2-2-hydroxyethoxy ethane, 5-hydroxy-3-oxapentylamine | |
2-(2-aminoethoxy)ethanol |
C4H11NO2 | |
GIAFURWZWWWBQT-UHFFFAOYSA-N | |
13578 | |
C(COCCO)N |
Specifications
929-06-6 | |
Yellow | |
218.0°C to 224.0°C | |
Authentic | |
C4H11NO2 | |
1.4585 to 1.4605 | |
1.06 | |
Solubility in water: miscible. Other solubilities: miscible with alcohol and aromatic hydrocarbons, practically immiscible with aliphatic hydrocarbons, and ethyl ether | |
C(COCCO)N | |
105.14 | |
105.14 | |
Crystalline Powder and/or Chunks |
-12.5°C | |
1.0600g/mL | |
127°C | |
97.5% min. (GC) | |
H2NCH2CH2OCH2CH2OH | |
04,III,642 | |
2-2-aminoethoxy ethanol, diglycolamine, 2-aminoethoxyethanol, ethanol, 2-2-aminoethoxy, diethylene glycol amine, 2-2-hydroxyethoxy ethylamine, diethylene glycol monoamine, 2-amino-2'-hydroxydiethyl ether, 1-amino-2-2-hydroxyethoxy ethane, 5-hydroxy-3-oxapentylamine | |
GIAFURWZWWWBQT-UHFFFAOYSA-N | |
2-(2-aminoethoxy)ethanol | |
13578 | |
98% | |
2-(2-Aminoethoxy)ethanol |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 213-195-4
- 2-(2-Aminoethoxy)ethanol
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only