Learn More
2,3,4-Tribromothiophene, 97%, Thermo Scientific™
£43.10 - £182.00
Chemical Identifiers
CAS | 3141-25-1 |
---|---|
Molecular Formula | C4HBr3S |
Molecular Weight (g/mol) | 320.82 |
MDL Number | MFCD00041210 |
InChI Key | ZDXQFDMBZUQHOM-UHFFFAOYSA-N |
PubChem CID | 76593 |
IUPAC Name | 2,3,4-tribromothiophene |
SMILES | BrC1=C(Br)C(Br)=CS1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11306726
|
Thermo Scientific Alfa Aesar
L03946.03 |
1 g | |||||||
11316726
|
Thermo Scientific Alfa Aesar
L03946.06 |
5 g |
£182.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
3141-25-1 | |
320.82 | |
ZDXQFDMBZUQHOM-UHFFFAOYSA-N | |
2,3,4-tribromothiophene |
C4HBr3S | |
MFCD00041210 | |
76593 | |
BrC1=C(Br)C(Br)=CS1 |
Specifications
3141-25-1 | |
150°C to 152°C (20 mmHg) | |
MFCD00041210 | |
111770 | |
ZDXQFDMBZUQHOM-UHFFFAOYSA-N | |
2,3,4-tribromothiophene | |
76593 | |
97% |
43°C to 45°C | |
C4HBr3S | |
UN2811 | |
tribromothiophene, thiophene, 2,3,4-tribromo, pubchem7738, acmc-1bnbq, 3,4,5-tribromothiophene, thiophene,2,3,4-tribromo, buttpark 29\09-41 | |
BrC1=C(Br)C(Br)=CS1 | |
320.82 | |
320.84 | |
2,3,4-Tribromothiophene |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 221-545-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures