missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Trifluorophenylacetic acid, 97%, Thermo Scientific™
£27.10 - £62.39
Chemical Identifiers
CAS | 132992-28-0 |
---|---|
Molecular Formula | C8H5F3O2 |
Molecular Weight (g/mol) | 190.121 |
MDL Number | MFCD00083547 |
InChI Key | HHVKNKDKNBABCU-UHFFFAOYSA-N |
Synonym | 2,3,5-trifluorophenylacetic acid, 2-2,3,5-trifluorophenyl acetic acid, 2,3,5-trifluorophenyl acetic acid, benzeneacetic acid, 2,3,5-trifluoro, pubchem4205, intermediates-zcf02025, acmc-1c03i, 2,3,5-trifluorobenzeneacetic acid, 2-2,3,5-trifluorophenyl aceticacid |
PubChem CID | 2777946 |
IUPAC Name | 2-(2,3,5-trifluorophenyl)acetic acid |
SMILES | C1=C(C=C(C(=C1F)F)CC(=O)O)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15483547
|
Thermo Scientific Alfa Aesar
H26826.MD |
250 mg |
£27.10
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11316969
|
Thermo Scientific Alfa Aesar
H26826.03 |
1 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
132992-28-0 | |
190.121 | |
HHVKNKDKNBABCU-UHFFFAOYSA-N | |
2777946 | |
C1=C(C=C(C(=C1F)F)CC(=O)O)F |
C8H5F3O2 | |
MFCD00083547 | |
2,3,5-trifluorophenylacetic acid, 2-2,3,5-trifluorophenyl acetic acid, 2,3,5-trifluorophenyl acetic acid, benzeneacetic acid, 2,3,5-trifluoro, pubchem4205, intermediates-zcf02025, acmc-1c03i, 2,3,5-trifluorobenzeneacetic acid, 2-2,3,5-trifluorophenyl aceticacid | |
2-(2,3,5-trifluorophenyl)acetic acid |
Specifications
132992-28-0 | |
MFCD00083547 | |
HHVKNKDKNBABCU-UHFFFAOYSA-N | |
2-(2,3,5-trifluorophenyl)acetic acid | |
2777946 | |
97% |
C8H5F3O2 | |
2,3,5-trifluorophenylacetic acid, 2-2,3,5-trifluorophenyl acetic acid, 2,3,5-trifluorophenyl acetic acid, benzeneacetic acid, 2,3,5-trifluoro, pubchem4205, intermediates-zcf02025, acmc-1c03i, 2,3,5-trifluorobenzeneacetic acid, 2-2,3,5-trifluorophenyl aceticacid | |
C1=C(C=C(C(=C1F)F)CC(=O)O)F | |
190.121 | |
190.12 | |
2,3,5-Trifluorophenylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures