missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromo-4-hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

£27.70 - £333.00

Chemical Identifiers

CAS 2973-75-3
Molecular Formula C8H6Br2O3
Molecular Weight (g/mol) 309.94
MDL Number MFCD00016978
InChI Key WKLKGSHBXNPUDU-UHFFFAOYSA-N
Synonym 5,6-dibromovanillin, acmc-1chm2, 2,3-dibromo-5-methoxy-4-hydroxybenzaldehyde, 2,3-bis bromanyl-5-methoxy-4-oxidanyl-benzaldehyde
PubChem CID 520452
IUPAC Name 2,3-dibromo-4-hydroxy-5-methoxybenzaldehyde
SMILES COC1=CC(C=O)=C(Br)C(Br)=C1O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11404637
View Documents
Thermo Scientific Alfa Aesar
B21140.03
1 g
£27.70
1g
Please to purchase this item. Need a web account? Register with us today!
11414637
View Documents
Thermo Scientific Alfa Aesar
B21140.06
5 g
£85.60
5g
Please to purchase this item. Need a web account? Register with us today!
11424637
View Documents
Thermo Scientific Alfa Aesar
B21140.14
25 g
£333.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2973-75-3
309.94
WKLKGSHBXNPUDU-UHFFFAOYSA-N
520452
COC1=CC(C=O)=C(Br)C(Br)=C1O
C8H6Br2O3
MFCD00016978
5,6-dibromovanillin, acmc-1chm2, 2,3-dibromo-5-methoxy-4-hydroxybenzaldehyde, 2,3-bis bromanyl-5-methoxy-4-oxidanyl-benzaldehyde
2,3-dibromo-4-hydroxy-5-methoxybenzaldehyde

Specifications

2973-75-3
C8H6Br2O3
1968576
5,6-dibromovanillin, acmc-1chm2, 2,3-dibromo-5-methoxy-4-hydroxybenzaldehyde, 2,3-bis bromanyl-5-methoxy-4-oxidanyl-benzaldehyde
COC1=CC(C=O)=C(Br)C(Br)=C1O
309.94
309.94
2,3-Dibromo-4-hydroxy-5-methoxybenzaldehyde
213°C to 217°C
MFCD00016978
Air Sensitive
WKLKGSHBXNPUDU-UHFFFAOYSA-N
2,3-dibromo-4-hydroxy-5-methoxybenzaldehyde
520452
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.