missing translation for 'onlineSavingsMsg'
Learn More
2,3-Dibromopropene, 80%, tech., stabilized, Thermo Scientific Chemicals Product Code.: 10645962

2,3-Dibromopropene, 80%, tech., stabilized, Thermo Scientific Chemicals

Product Code. 10645962
10 mL, Glass bottle
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10645962

Brand: Thermo Scientific Acros 147740100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-31-5
Molecular Formula C3H4Br2
Molecular Weight (g/mol) 199.87
MDL Number MFCD00000211
InChI Key YMFWYDYJHRGGPF-UHFFFAOYSA-N
Synonym 2,3-dibromopropene, 2,3-dibromo-1-propene, 1-propene, 2,3-dibromo, 2-bromoallyl bromide, 2,3-dibromopropylene, 1,2-dibromo-2-propene, alpha-epidibromohydrin, propene, 2,3-dibromo, unii-8dne7k7z6p, ccris 6712Show MoreShow Less
PubChem CID 10552
IUPAC Name 2,3-dibromoprop-1-ene
SMILES BrCC(Br)=C

Specifications

CAS Min % 75.0
CAS Max % 100.0
Color Yellow to Brown
Density 2.1300g/mL
Boiling Point 42°C to 44°C (17.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 75% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2C(Br)=CH2
Refractive Index 1.5370 to 1.547
Quantity 10 mL
Beilstein 01, 201
Merck Index 15, 3030
Specific Gravity 2.13
Solubility Information Solubility in water: insoluble
Formula Weight 199.87
Percent Purity 80%
Grade Technical
Physical Form Liquid
Chemical Name or Material 2, 3-Dibromopropene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453564 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number147740100,147740500,147742500
Lot NumberA0461253 Certificate TypeCertificate of Analysis Date26/04/2024 Catalog Number147740100,147740500,147742500
Lot NumberA0453564 Certificate TypeCertificate of Analysis Date14/12/2023 Catalog Number147740100,147740500,147742500
Lot NumberA0443897 Certificate TypeCertificate of Analysis Date06/02/2023 Catalog Number147740100,147740500,147742500
Lot NumberA0400789 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number147740100,147740500,147742500
compliance-icons
Product Identifier
  • 2,3-Dibromopropene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3-Dibromopropene, 80%, tech., stabilized, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.