missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydro-1,4-benzodioxine-6-carbaldehyde 97%, Thermo Scientific™
£8.10 - £20.70
Chemical Identifiers
CAS | 29668-44-8 |
---|---|
Molecular Formula | C9H8O3 |
Molecular Weight (g/mol) | 164.16 |
InChI Key | CWKXDPPQCVWXAG-UHFFFAOYSA-N |
Synonym | 1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehyde |
PubChem CID | 248127 |
IUPAC Name | 2,3-dihydro-1,4-benzodioxine-6-carbaldehyde |
SMILES | C1COC2=C(O1)C=CC(=C2)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10181302
|
Thermo Scientific Maybridge
CC01304DA |
1 g |
£8.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10177814
|
Thermo Scientific Maybridge
CC01304DE |
5 g |
£20.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
29668-44-8 | |
164.16 | |
1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehyde | |
2,3-dihydro-1,4-benzodioxine-6-carbaldehyde |
C9H8O3 | |
CWKXDPPQCVWXAG-UHFFFAOYSA-N | |
248127 | |
C1COC2=C(O1)C=CC(=C2)C=O |
Specifications
29668-44-8 | |
Amber glass bottle | |
1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehyde | |
C1COC2=C(O1)C=CC(=C2)C=O | |
164.16 | |
164.16 | |
2, 3-Dihydro-1, 4-benzodioxine-6-carbaldehyde |
97% | |
C9H8O3 | |
CWKXDPPQCVWXAG-UHFFFAOYSA-N | |
2,3-dihydro-1,4-benzodioxine-6-carbaldehyde | |
248127 | |
97% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2,3-Dihydro-1,4-benzodioxine-6-carbaldehyde 97%, Thermo Scientific™