missing translation for 'onlineSavingsMsg'
Learn More
2,3-Dihydro-1,4-benzodioxine-6-carbaldehyde 97%, Thermo Scientific™ Product Code.: 10177814

2,3-Dihydro-1,4-benzodioxine-6-carbaldehyde 97%, Thermo Scientific™

Product Code. 10177814
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 10177814

Brand: Thermo Scientific Maybridge CC01304DE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 29668-44-8
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
InChI Key CWKXDPPQCVWXAG-UHFFFAOYSA-N
Synonym 1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehydeShow MoreShow Less
PubChem CID 248127
IUPAC Name 2,3-dihydro-1,4-benzodioxine-6-carbaldehyde
SMILES C1COC2=C(O1)C=CC(=C2)C=O

Specifications

Assay Percent Range 97%
Quantity 5 g
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-6-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3-Dihydro-1,4-benzodioxine-6-carbaldehyde 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.