missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals
£21.30 - £330.00
Chemical Identifiers
CAS | 40972-86-9 |
---|---|
Molecular Formula | C8H11BO4 |
Molecular Weight (g/mol) | 181.98 |
MDL Number | MFCD02683112 |
InChI Key | VREWSCMOGIXMDQ-UHFFFAOYSA-N |
Synonym | 2,3-dimethoxybenzeneboronic acid, 2,3-dimethoxyphenyl boronic acid, boronic acid, 2,3-dimethoxyphenyl, 2,3-dimethoxyphenylboronicacid, 3-boronoveratrole, pubchem14109, acmc-209jhv, ksc235o1n, 2,3-dimethoxy-phenylboronic acid |
PubChem CID | 5156491 |
IUPAC Name | (2,3-dimethoxyphenyl)boronic acid |
SMILES | COC1=CC=CC(B(O)O)=C1OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11491127
|
Thermo Scientific Alfa Aesar
B24125.03 |
1 g | |||||||
11401137
|
Thermo Scientific Alfa Aesar
B24125.06 |
5 g |
£73.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11411137
|
Thermo Scientific Alfa Aesar
B24125.14 |
25 g |
£330.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
40972-86-9 | |
181.98 | |
VREWSCMOGIXMDQ-UHFFFAOYSA-N | |
5156491 | |
COC1=CC=CC(B(O)O)=C1OC |
C8H11BO4 | |
MFCD02683112 | |
2,3-dimethoxybenzeneboronic acid, 2,3-dimethoxyphenyl boronic acid, boronic acid, 2,3-dimethoxyphenyl, 2,3-dimethoxyphenylboronicacid, 3-boronoveratrole, pubchem14109, acmc-209jhv, ksc235o1n, 2,3-dimethoxy-phenylboronic acid | |
(2,3-dimethoxyphenyl)boronic acid |
Specifications
40972-86-9 | |
C8H11BO4 | |
3051295 | |
VREWSCMOGIXMDQ-UHFFFAOYSA-N | |
(2,3-dimethoxyphenyl)boronic acid | |
5156491 | |
98% |
67°C to 71°C | |
MFCD02683112 | |
2,3-dimethoxybenzeneboronic acid, 2,3-dimethoxyphenyl boronic acid, boronic acid, 2,3-dimethoxyphenyl, 2,3-dimethoxyphenylboronicacid, 3-boronoveratrole, pubchem14109, acmc-209jhv, ksc235o1n, 2,3-dimethoxy-phenylboronic acid | |
COC1=CC=CC(B(O)O)=C1OC | |
181.98 | |
181.98 | |
2,3-Dimethoxybenzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only