Learn More
2,4-Dimethylaniline, 99%, Thermo Scientific Chemicals
£20.60 - £117.00
Chemical Identifiers
CAS | 95-68-1 |
---|---|
Molecular Formula | C8H11N |
Molecular Weight (g/mol) | 121.18 |
MDL Number | MFCD00007738 |
InChI Key | CZZZABOKJQXEBO-UHFFFAOYSA-N |
Synonym | 2,4-xylidine, 2,4-dimethyl aniline, m-xylidine, m-4-xylidine, 2,4-dimethylphenylamine, benzenamine, 2,4-dimethyl, 2-methyl-p-toluidine, 4-methyl-o-toluidine, 1-amino-2,4-dimethylbenzene, 4-amino-1,3-xylene |
PubChem CID | 7250 |
ChEBI | CHEBI:27840 |
IUPAC Name | 2,4-dimethylaniline |
SMILES | CC1=CC(=C(C=C1)N)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10736191
|
Thermo Scientific Acros
|
100 g | Glass bottle |
£20.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10031520
|
Thermo Scientific Acros
|
1 kg | Glass bottle |
£117.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-68-1 | |
121.18 | |
CZZZABOKJQXEBO-UHFFFAOYSA-N | |
7250 | |
2,4-dimethylaniline |
C8H11N | |
MFCD00007738 | |
2,4-xylidine, 2,4-dimethyl aniline, m-xylidine, m-4-xylidine, 2,4-dimethylphenylamine, benzenamine, 2,4-dimethyl, 2-methyl-p-toluidine, 4-methyl-o-toluidine, 1-amino-2,4-dimethylbenzene, 4-amino-1,3-xylene | |
CHEBI:27840 | |
CC1=CC(=C(C=C1)N)C |
Specifications
16.0°C | |
98.5 | |
0.9800g/mL | |
Authentic | |
1.5580 to 1.5600 | |
0.98 | |
(CH3)2C6H3NH2 | |
12, 1111 | |
2,4-xylidine, 2,4-dimethyl aniline, m-xylidine, m-4-xylidine, 2,4-dimethylphenylamine, benzenamine, 2,4-dimethyl, 2-methyl-p-toluidine, 4-methyl-o-toluidine, 1-amino-2,4-dimethylbenzene, 4-amino-1,3-xylene | |
CZZZABOKJQXEBO-UHFFFAOYSA-N | |
2,4-dimethylaniline | |
7250 | |
121.18 | |
2,4-Dimethylaniline, 99% |
95-68-1 | |
100.0 | |
218.0°C | |
Liquid | |
98.5% min. (GC) | |
C8H11N | |
MFCD00007738 | |
15, 10282 | |
Solubility in water: 5g/L (20°C). Other solubilities: soluble in ethanol, ether and benzene | |
CC1=CC(=C(C=C1)N)C | |
121.18 | |
CHEBI:27840 | |
99% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER o
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-440-0
missing translation for 'rtecsNumber' : ZE8925000
missing translation for 'tsca' : TSCA
Recommended Storage : Product darkens if exposed to light and air
- 2,4-Dimethylaniline
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only