Learn More
2,5-Di-tert-butylhydroquinone, 99%, Thermo Scientific Chemicals
£62.30 - £134.00
Chemical Identifiers
CAS | 88-58-4 |
---|---|
Molecular Formula | C14H22O2 |
Molecular Weight (g/mol) | 222.33 |
MDL Number | MFCD00008825 |
InChI Key | JZODKRWQWUWGCD-UHFFFAOYSA-N |
Synonym | 2,5-di-tert-butylhydroquinone, 2,5-di-tert-butylbenzene-1,4-diol, dibug, dtbhq, dybug, santovar o, di-t-butylhydroquinone, 2,5-di-tert-butylquinol, nonflex alba, 2,5-di-t-butylhydroquinone |
PubChem CID | 2374 |
ChEBI | CHEBI:41094 |
IUPAC Name | 2,5-ditert-butylbenzene-1,4-diol |
SMILES | CC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10215960
|
Thermo Scientific Acros
150840010 |
1 kg | Plastic bottle |
£62.30
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10404555
|
Thermo Scientific Acros
150840025 |
2.5 kg | Plastic bottle |
£134.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
88-58-4 | |
222.33 | |
JZODKRWQWUWGCD-UHFFFAOYSA-N | |
2374 | |
2,5-ditert-butylbenzene-1,4-diol |
C14H22O2 | |
MFCD00008825 | |
2,5-di-tert-butylhydroquinone, 2,5-di-tert-butylbenzene-1,4-diol, dibug, dtbhq, dybug, santovar o, di-t-butylhydroquinone, 2,5-di-tert-butylquinol, nonflex alba, 2,5-di-t-butylhydroquinone | |
CHEBI:41094 | |
CC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)C |
Specifications
88-58-4 | |
100.0 | |
Beige to Beige-Pink | |
216°C | |
98.5% min. (HPLC) | |
MFCD00008825 | |
Solubility in water: insoluble in water. Other solubilities: insoluble in aqueous alkali,soluble in acetone,alcohol and benzene,practically insoluble in toluene and hexane | |
CC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)C | |
222.33 | |
CHEBI:41094 | |
99% | |
2, 5-Di-tert-butylhydroquinone |
98.5 | |
212°C to 218°C | |
216g/mL | |
Authentic | |
C14H22O2 | |
2,5-di-tert-butylhydroquinone, 2,5-di-tert-butylbenzene-1,4-diol, dibug, dtbhq, dybug, santovar o, di-t-butylhydroquinone, 2,5-di-tert-butylquinol, nonflex alba, 2,5-di-t-butylhydroquinone | |
JZODKRWQWUWGCD-UHFFFAOYSA-N | |
2,5-ditert-butylbenzene-1,4-diol | |
2374 | |
222.33 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 201-841-8
RUO – Research Use Only