Learn More
2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane, 92%, Thermo Scientific Chemicals
£15.78 - £35.78
Chemical Identifiers
CAS | 78-63-7 |
---|---|
Molecular Formula | C16H34O4 |
Molecular Weight (g/mol) | 290.44 |
InChI Key | DMWVYCCGCQPJEA-UHFFFAOYSA-N |
Synonym | varox, 2,5-dimethyl-2,5-di tert-butylperoxy hexane, 2,5-bis tert-butylperoxy-2,5-dimethylhexane, triqanox xq 8, kayahexa ad 40c, lupersol 101, luperco 101xl, luperox 101, perhexa 3m40, varox 50 |
PubChem CID | 6545 |
IUPAC Name | 2,5-bis(tert-butylperoxy)-2,5-dimethylhexane |
SMILES | CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10623512
|
Thermo Scientific
349900050 |
5 g | Plastic bottle |
£15.78
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10153191
|
Thermo Scientific Acros
349901000 |
100 g | Plastic bottle |
£35.78
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiatorsChemical Identifiers
78-63-7 | |
290.44 | |
varox, 2,5-dimethyl-2,5-di tert-butylperoxy hexane, 2,5-bis tert-butylperoxy-2,5-dimethylhexane, triqanox xq 8, kayahexa ad 40c, lupersol 101, luperco 101xl, luperox 101, perhexa 3m40, varox 50 | |
2,5-bis(tert-butylperoxy)-2,5-dimethylhexane |
C16H34O4 | |
DMWVYCCGCQPJEA-UHFFFAOYSA-N | |
6545 | |
CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C |
Specifications
78-63-7 | |
0.8700g/mL | |
68°C | |
C16H34O4 | |
0.87 | |
Solubility in water: immiscible. Other solubilities: soluble in organic solvents,soluble in alcohol | |
CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C | |
290.44 | |
6.4 mPa.s (20°C) | |
92% |
6.0°C | |
55.0°C to 57.0°C (7.0 mmHg) | |
90% min. (GC) | |
1.4200 to 1.4240 | |
varox, 2,5-dimethyl-2,5-di tert-butylperoxy hexane, 2,5-bis tert-butylperoxy-2,5-dimethylhexane, triqanox xq 8, kayahexa ad 40c, lupersol 101, luperco 101xl, luperox 101, perhexa 3m40, varox 50 | |
DMWVYCCGCQPJEA-UHFFFAOYSA-N | |
2,5-bis(tert-butylperoxy)-2,5-dimethylhexane | |
6545 | |
290.44 | |
2, 5-Dimethyl-2, 5-di(tert-butylperoxy)hexane |
Safety and Handling
GHS H Statement
Causes skin irritation.
Heating may cause a fire.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep/Store away from clothing/ combustible materials.
Store in a well-v
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-128-1
- 2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane
Signal Word
- Danger
Hazard Category
- Organic peroxide Type C & D
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H242-Heating may cause a fire.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P403+P235-Store in a well-ventilated place. Keep cool.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.