Learn More
2,6-Di-tert-butyl-4-methoxyphenol, 97%, Thermo Scientific Chemicals
£33.67 - £92.26
Chemical Identifiers
CAS | 489-01-0 |
---|---|
Molecular Formula | C15H24O2 |
Molecular Weight (g/mol) | 236.35 |
MDL Number | MFCD00008824 |
InChI Key | SLUKQUGVTITNSY-UHFFFAOYSA-N |
Synonym | 2,6-di-tert-butyl-4-methoxyphenol, 3,5-di-tert-butyl-4-hydroxyanisole, topanol 354, phenol, 2,6-bis 1,1-dimethylethyl-4-methoxy, 3,5-di-t-butyl-4-hydroxyanisole, unii-616072tmxy, ccris 5219, 2,6-bis 1,1-dimethylethyl-4-methoxyphenol, phenol, 2,6-di-tert-butyl-4-methoxy, chembl98253 |
PubChem CID | 10269 |
IUPAC Name | 2,6-ditert-butyl-4-methoxyphenol |
SMILES | CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10741911
|
Thermo Scientific Acros
294160250 |
25 g | Glass bottle |
£33.67
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10339250
|
Thermo Scientific Acros
294161000 |
100 g | Glass bottle |
£92.26
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
489-01-0 | |
236.35 | |
SLUKQUGVTITNSY-UHFFFAOYSA-N | |
10269 | |
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)OC |
C15H24O2 | |
MFCD00008824 | |
2,6-di-tert-butyl-4-methoxyphenol, 3,5-di-tert-butyl-4-hydroxyanisole, topanol 354, phenol, 2,6-bis 1,1-dimethylethyl-4-methoxy, 3,5-di-t-butyl-4-hydroxyanisole, unii-616072tmxy, ccris 5219, 2,6-bis 1,1-dimethylethyl-4-methoxyphenol, phenol, 2,6-di-tert-butyl-4-methoxy, chembl98253 | |
2,6-ditert-butyl-4-methoxyphenol |
Specifications
489-01-0 | |
100.0 | |
Authentic | |
C15H24O2 | |
16,110; 16,58 | |
Solubility in water: practically insoluble | |
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)OC | |
236.35 | |
236.35 | |
Liquid |
96.0 | |
103°C to 106°C | |
96% min. (GC) | |
MFCD00008824 | |
2,6-di-tert-butyl-4-methoxyphenol, 3,5-di-tert-butyl-4-hydroxyanisole, topanol 354, phenol, 2,6-bis 1,1-dimethylethyl-4-methoxy, 3,5-di-t-butyl-4-hydroxyanisole, unii-616072tmxy, ccris 5219, 2,6-bis 1,1-dimethylethyl-4-methoxyphenol, phenol, 2,6-di-tert-butyl-4-methoxy, chembl98253 | |
SLUKQUGVTITNSY-UHFFFAOYSA-N | |
2,6-ditert-butyl-4-methoxyphenol | |
10269 | |
97% | |
2, 6-Di-tert-butyl-4-methoxyphenol, 97% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 207-693-0
missing translation for 'rtecsNumber' : SJ7785668
missing translation for 'tsca' : TSCA
- 2,6-Di-tert-butyl-4-methoxyphenol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only