Learn More
2,6-Dibromopyrazine, 95%, Thermo Scientific Chemicals
£63.50 - £241.00
Chemical Identifiers
CAS | 23229-25-6 |
---|---|
Molecular Formula | C4H2Br2N2 |
Molecular Weight (g/mol) | 237.88 |
MDL Number | MFCD09834804 |
InChI Key | JXKQTRCEKQCAGH-UHFFFAOYSA-N |
Synonym | pyrazine, 2,6-dibromo, 2,6-dibromo-pyrazine, acmc-1cj6s, ksc201c7f |
PubChem CID | 20361700 |
IUPAC Name | 2,6-dibromopyrazine |
SMILES | BrC1=CN=CC(Br)=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15478018
|
Thermo Scientific Alfa Aesar
H61468.03 |
1 g |
£63.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15488018
|
Thermo Scientific Alfa Aesar
H61468.06 |
5 g |
£241.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,6-Dibromopyrazine is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Dibromopyrazine is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Storage temperature 2 - 4°Cand Keep container tightly closed in a dry and well-ventilated place.
Identifiants chimiques
23229-25-6 | |
237.88 | |
JXKQTRCEKQCAGH-UHFFFAOYSA-N | |
20361700 | |
BrC1=CN=CC(Br)=N1 |
C4H2Br2N2 | |
MFCD09834804 | |
pyrazine, 2,6-dibromo, 2,6-dibromo-pyrazine, acmc-1cj6s, ksc201c7f | |
2,6-dibromopyrazine |
Spécification
23229-25-6 | |
C4H2Br2N2 | |
pyrazine, 2,6-dibromo, 2,6-dibromo-pyrazine, acmc-1cj6s, ksc201c7f | |
JXKQTRCEKQCAGH-UHFFFAOYSA-N | |
2,6-dibromopyrazine | |
20361700 | |
95% | |
2,6-Dibromopyrazine |
White to Brown | |
MFCD09834804 | |
Slightly soluble in water. | |
BrC1=CN=CC(Br)=N1 | |
237.88 | |
237.88 | |
Crystalline Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
missing translation for 'storageNote' : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only