missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2,6-Dichlorobenzyl)triphenylphosphonium chloride, 98%, Thermo Scientific™
£20.70 - £214.00
Chemical Identifiers
CAS | 18583-54-5 |
---|---|
Molecular Formula | C25H20Cl3P |
Molecular Weight (g/mol) | 457.76 |
MDL Number | MFCD00061392 |
InChI Key | NBWPWZJEEQWREI-UHFFFAOYSA-M |
Synonym | 2,6-dichlorobenzyl triphenylphosphonium chloride, 2,6-dichlorophenyl methyl triphenylphosphanium chloride, acmc-20ao99, 2,6-dichlorobenzyltriphenylphosphoniumchloride, 2,6-dichlorophenyl methyl-triphenyl-phosphonium chloride, 2,6-dichlorophenyl methyl triphenyl phosphanium chloride, phosphonium, 2,6-dichlorophenyl methyl triphenyl-, chloride 1:1 |
PubChem CID | 57370833 |
IUPAC Name | (2,6-dichlorophenyl)methyl-triphenylphosphanium;chloride |
SMILES | [Cl-].ClC1=CC=CC(Cl)=C1C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificatori chimici
18583-54-5 | |
457.76 | |
NBWPWZJEEQWREI-UHFFFAOYSA-M | |
57370833 | |
[Cl-].ClC1=CC=CC(Cl)=C1C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C25H20Cl3P | |
MFCD00061392 | |
2,6-dichlorobenzyl triphenylphosphonium chloride, 2,6-dichlorophenyl methyl triphenylphosphanium chloride, acmc-20ao99, 2,6-dichlorobenzyltriphenylphosphoniumchloride, 2,6-dichlorophenyl methyl-triphenyl-phosphonium chloride, 2,6-dichlorophenyl methyl triphenyl phosphanium chloride, phosphonium, 2,6-dichlorophenyl methyl triphenyl-, chloride 1:1 | |
(2,6-dichlorophenyl)methyl-triphenylphosphanium;chloride |
Specifica
18583-54-5 | |
MFCD00061392 | |
NBWPWZJEEQWREI-UHFFFAOYSA-M | |
(2,6-dichlorophenyl)methyl-triphenylphosphanium;chloride | |
57370833 | |
98% |
C25H20Cl3P | |
2,6-dichlorobenzyl triphenylphosphonium chloride, 2,6-dichlorophenyl methyl triphenylphosphanium chloride, acmc-20ao99, 2,6-dichlorobenzyltriphenylphosphoniumchloride, 2,6-dichlorophenyl methyl-triphenyl-phosphonium chloride, 2,6-dichlorophenyl methyl triphenyl phosphanium chloride, phosphonium, 2,6-dichlorophenyl methyl triphenyl-, chloride 1:1 | |
[Cl-].ClC1=CC=CC(Cl)=C1C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
457.76 | |
457.78 | |
(2,6-Dichlorobenzyl)triphenylphosphonium chloride |
Sicurezza e movimentazione
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures