Learn More
2,6-Diethylaniline, 98%, Thermo Scientific Chemicals
£32.90 - £93.50
Chemical Identifiers
CAS | 579-66-8 |
---|---|
Molecular Formula | C10H15N |
Molecular Weight (g/mol) | 149.24 |
MDL Number | MFCD00007753 |
InChI Key | FOYHNROGBXVLLX-UHFFFAOYSA-N |
Synonym | benzenamine, 2,6-diethyl, 2,6-diethylbenzenamine, aniline, 2,6-diethyl, 2,6-diethyl aniline, 2-amino-1,3-diethylbenzene, 2,6-diethyl-aniline, ccris 2688, benzamine, 2,6-diethyl, 2,6 diethylaniline, 2,6-diethylphenylamine |
PubChem CID | 11369 |
ChEBI | CHEBI:27880 |
IUPAC Name | 2,6-diethylaniline |
SMILES | CCC1=C(C(=CC=C1)CC)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10694832
|
Thermo Scientific Acros
165222500 |
250 mL | Glass bottle |
£32.90
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10317680
|
Thermo Scientific Acros
165220010 |
1 L | Glass bottle |
£93.50
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
579-66-8 | |
149.24 | |
FOYHNROGBXVLLX-UHFFFAOYSA-N | |
11369 | |
2,6-diethylaniline |
C10H15N | |
MFCD00007753 | |
benzenamine, 2,6-diethyl, 2,6-diethylbenzenamine, aniline, 2,6-diethyl, 2,6-diethyl aniline, 2-amino-1,3-diethylbenzene, 2,6-diethyl-aniline, ccris 2688, benzamine, 2,6-diethyl, 2,6 diethylaniline, 2,6-diethylphenylamine | |
CHEBI:27880 | |
CCC1=C(C(=CC=C1)CC)N |
Specifications
579-66-8 | |
100.0 | |
Brown-Red to Yellow | |
243°C | |
Authentic | |
C10H15N | |
1.5446 to 1.5466 | |
12,164 | |
benzenamine, 2,6-diethyl, 2,6-diethylbenzenamine, aniline, 2,6-diethyl, 2,6-diethyl aniline, 2-amino-1,3-diethylbenzene, 2,6-diethyl-aniline, ccris 2688, benzamine, 2,6-diethyl, 2,6 diethylaniline, 2,6-diethylphenylamine | |
CCC1=C(C(=CC=C1)CC)N | |
149.24 | |
CHEBI:27880 | |
98% | |
2, 6-Diethylaniline, 98% |
97.5 | |
3°C to 4°C | |
0.9000g/mL | |
123°C | |
97.5% min. (GC) | |
(C2H5)2C6H3NH2 | |
MFCD00007753 | |
0.9 | |
FOYHNROGBXVLLX-UHFFFAOYSA-N | |
2,6-diethylaniline | |
11369 | |
149.24 | |
Liquid |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-445-7
missing translation for 'rtecsNumber' : BX3500000
missing translation for 'tsca' : TSCA
Recommended Storage : substance rapidly darkens on exposure to light and air
RUO – Research Use Only