missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-3-methylphenylacetic acid, 97%, Thermo Scientific™

£47.02 - £129.00

Chemical Identifiers

CAS 261943-97-9
Molecular Formula C9H8F2O2
Molecular Weight (g/mol) 186.158
MDL Number MFCD01631338
InChI Key XMNMLOQKAUVILT-UHFFFAOYSA-N
Synonym 2,6-difluoro-3-methylphenylacetic acid, 2-2,6-difluoro-3-methylphenyl acetic acid, 2,6-difluoro-3-methylphenyl acetic acid, acmc-1ce69, benzeneacetic acid,2,6-difluoro-3-methyl
PubChem CID 2774155
IUPAC Name 2-(2,6-difluoro-3-methylphenyl)acetic acid
SMILES CC1=C(C(=C(C=C1)F)CC(=O)O)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15453457
View Documents
Thermo Scientific Alfa Aesar
H26381.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11491310
View Documents
Thermo Scientific Alfa Aesar
H26381.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

261943-97-9
186.158
XMNMLOQKAUVILT-UHFFFAOYSA-N
2774155
CC1=C(C(=C(C=C1)F)CC(=O)O)F
C9H8F2O2
MFCD01631338
2,6-difluoro-3-methylphenylacetic acid, 2-2,6-difluoro-3-methylphenyl acetic acid, 2,6-difluoro-3-methylphenyl acetic acid, acmc-1ce69, benzeneacetic acid,2,6-difluoro-3-methyl
2-(2,6-difluoro-3-methylphenyl)acetic acid

Specifications

261943-97-9
MFCD01631338
XMNMLOQKAUVILT-UHFFFAOYSA-N
2-(2,6-difluoro-3-methylphenyl)acetic acid
2774155
97%
C9H8F2O2
2,6-difluoro-3-methylphenylacetic acid, 2-2,6-difluoro-3-methylphenyl acetic acid, 2,6-difluoro-3-methylphenyl acetic acid, acmc-1ce69, benzeneacetic acid,2,6-difluoro-3-methyl
CC1=C(C(=C(C=C1)F)CC(=O)O)F
186.158
186.16
2,6-Difluoro-3-methylphenylacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications