Learn More
2',6'-Difluoroacetophenone, 98%, Thermo Scientific Chemicals
£27.60 - £238.00
Chemical Identifiers
CAS | 13670-99-0 |
---|---|
Molecular Formula | C8H6F2O |
Molecular Weight (g/mol) | 156.13 |
MDL Number | MFCD00000328 |
InChI Key | VGIIILXIQLXVLC-UHFFFAOYSA-N |
Synonym | 2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054 |
PubChem CID | 83643 |
SMILES | CC(=O)C1=C(F)C=CC=C1F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11409562
|
Thermo Scientific Alfa Aesar
A10220.03 |
1 g |
£27.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419562
|
Thermo Scientific Alfa Aesar
A10220.06 |
5 g |
£72.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11429562
|
Thermo Scientific Alfa Aesar
A10220.14 |
25 g |
£238.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2?,6?-Difluoroacetophenone was used in the synthesis of 2-amino-4-alkyl- and 2-amino-4-arylquinazolines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′,6′-Difluoroacetophenone was used in the synthesis of 2-amino-4-alkyl- and 2-amino-4-arylquinazolines.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
Chemical Identifiers
13670-99-0 | |
156.13 | |
VGIIILXIQLXVLC-UHFFFAOYSA-N | |
83643 |
C8H6F2O | |
MFCD00000328 | |
2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054 | |
CC(=O)C1=C(F)C=CC=C1F |
Specifications
13670-99-0 | |
76°C to 79°C (15 mmHg) | |
C8H6F2O | |
MFCD00000328 | |
2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054 | |
VGIIILXIQLXVLC-UHFFFAOYSA-N | |
1-(2,6-difluorophenyl)ethan-1-one | |
83643 | |
98% |
1.195 | |
76°C (168°F) | |
1.481 | |
2046068 | |
Slightly soluble in water. | |
CC(=O)C1=C(F)C=CC=C1F | |
156.13 | |
156.13 | |
2',6'-Difluoroacetophenone |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319-H335
missing translation for 'einecsNumber' : 237-151-9
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only