missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-([7-(acetylamino)-6-(benzyloxy)-2-phenylperhydropyrano[3,2-d][1,3]dioxin-8-yl]oxy)propanoic acid, Thermo Scientific™
£59.80 - £391.00
Chemical Identifiers
CAS | 499104-69-7 |
---|---|
Molecular Formula | C25H29NO8 |
Molecular Weight (g/mol) | 471.506 |
InChI Key | JPPMVSNCFXDOJX-UHFFFAOYSA-N |
Synonym | 2-7-acetylamino-6-benzyloxy-2-phenylperhydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2r-2-4ar,6s,7r,8r,8as-7-acetamido-6-benzyloxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2-6-benzyloxy-7-acetamido-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, benzyl n-acetyl-4,6-o-benzylidenemuramic acid, 2-7-acetamido-6-benzyloxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2-7-acetamido-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid |
PubChem CID | 3843768 |
IUPAC Name | 2-[(7-acetamido-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-8-yl)oxy]propanoic acid |
SMILES | CC(C(=O)O)OC1C(C(OC2C1OC(OC2)C3=CC=CC=C3)OCC4=CC=CC=C4)NC(=O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10681055
|
Thermo Scientific Maybridge
RH01183CB |
250 mg | Amber Glass Bottle |
£59.80
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10176902
|
Thermo Scientific Maybridge
RH01183DA |
1 g | Amber Glass Bottle |
£143.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10490574
|
Thermo Scientific Maybridge
RH01183DE |
5 g | Amber glass bottle |
£391.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
499104-69-7 | |
471.506 | |
2-7-acetylamino-6-benzyloxy-2-phenylperhydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2r-2-4ar,6s,7r,8r,8as-7-acetamido-6-benzyloxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2-6-benzyloxy-7-acetamido-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, benzyl n-acetyl-4,6-o-benzylidenemuramic acid, 2-7-acetamido-6-benzyloxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2-7-acetamido-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid | |
2-[(7-acetamido-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-8-yl)oxy]propanoic acid |
C25H29NO8 | |
JPPMVSNCFXDOJX-UHFFFAOYSA-N | |
3843768 | |
CC(C(=O)O)OC1C(C(OC2C1OC(OC2)C3=CC=CC=C3)OCC4=CC=CC=C4)NC(=O)C |
Specifications
499104-69-7 | |
C25H29NO8 | |
JPPMVSNCFXDOJX-UHFFFAOYSA-N | |
2-[(7-acetamido-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-8-yl)oxy]propanoic acid | |
3843768 | |
97% |
97% | |
2-7-acetylamino-6-benzyloxy-2-phenylperhydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2r-2-4ar,6s,7r,8r,8as-7-acetamido-6-benzyloxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2-6-benzyloxy-7-acetamido-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, benzyl n-acetyl-4,6-o-benzylidenemuramic acid, 2-7-acetamido-6-benzyloxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2-7-acetamido-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid | |
CC(C(=O)O)OC1C(C(OC2C1OC(OC2)C3=CC=CC=C3)OCC4=CC=CC=C4)NC(=O)C | |
471.506 | |
471.5 | |
2-{[7-(Acetylamino)-6-(benzyloxy)-2-phenylperhydropyrano[3, 2-d][1, 3]dioxin-8-yl]oxy}propanoic acid |