missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylbenzoic acid, 99%, Thermo Scientific Chemicals

£23.10 - £263.00

Chemical Identifiers

CAS 577-56-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00002475
InChI Key QDAWXRKTSATEOP-UHFFFAOYSA-N
Synonym o-acetylbenzoic acid, benzoic acid, 2-acetyl, 2-acetylbenzoicacid, acetophenone-2-carboxylic acid, 2-acetyl-benzoic acid, benzoic acid, acetyl, 3-hydroxy-3-methylphthalide, 2'-acetophenonecarboxylic acid, acetophenone-2'-carboxylic acid, methyl phenyl ketone-o-carboxylic acid
PubChem CID 68474
IUPAC Name 2-acetylbenzoic acid
SMILES CC(=O)C1=CC=CC=C1C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10122320
View Documents
Thermo Scientific Acros
102470050
5 g Glass bottle
£23.10
5g
Please to purchase this item. Need a web account? Register with us today!
10428050
View Documents
Thermo Scientific Acros
102470250
25 g Glass bottle
£72.80
25g
Please to purchase this item. Need a web account? Register with us today!
10581364
View Documents
Thermo Scientific Acros
102471000
100 g Glass bottle
£263.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

577-56-0
164.16
QDAWXRKTSATEOP-UHFFFAOYSA-N
68474
CC(=O)C1=CC=CC=C1C(O)=O
C9H8O3
MFCD00002475
o-acetylbenzoic acid, benzoic acid, 2-acetyl, 2-acetylbenzoicacid, acetophenone-2-carboxylic acid, 2-acetyl-benzoic acid, benzoic acid, acetyl, 3-hydroxy-3-methylphthalide, 2'-acetophenonecarboxylic acid, acetophenone-2'-carboxylic acid, methyl phenyl ketone-o-carboxylic acid
2-acetylbenzoic acid

Specifications

577-56-0
100.0
Yellow to Orange
99%
CH3COC6H4COOH
10, 690
QDAWXRKTSATEOP-UHFFFAOYSA-N
2-acetylbenzoic acid
68474
99%
2-Acetylbenzoic acid
98.5
115°C to 118°C
Authentic
C9H8O3
MFCD00002475
o-acetylbenzoic acid, benzoic acid, 2-acetyl, 2-acetylbenzoicacid, acetophenone-2-carboxylic acid, 2-acetyl-benzoic acid, benzoic acid, acetyl, 3-hydroxy-3-methylphthalide, 2'-acetophenonecarboxylic acid, acetophenone-2'-carboxylic acid, methyl phenyl ketone-o-carboxylic acid
CC(=O)C1=CC=CC=C1C(O)=O
164.16
164.16
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 209-413-2

Product Identifier
  • 2-Acetylbenzoic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Certificates

RUO – Research Use Only