Learn More
2-Acetylthiophene, 99%, Thermo Scientific Chemicals
£38.50 - £684.00
Chemical Identifiers
CAS | 88-15-3 |
---|---|
Molecular Formula | C6H6OS |
Molecular Weight (g/mol) | 126.173 |
MDL Number | MFCD00005442 |
InChI Key | WYJOVVXUZNRJQY-UHFFFAOYSA-N |
Synonym | 2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone |
PubChem CID | 6920 |
IUPAC Name | 1-thiophen-2-ylethanone |
SMILES | CC(=O)C1=CC=CS1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11444364
|
Thermo Scientific Alfa Aesar
A13062.22 |
100 g |
£38.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454364
|
Thermo Scientific Alfa Aesar
A13062.36 |
500 g |
£159.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11464364
|
Thermo Scientific Alfa Aesar
A13062.0E |
2500 g |
£684.00
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Acetylthiophene is used as an intermediate in the manufacturing of drugs like Tiamonium Iodide, Suprofan, Stepronin, Tenonitrozole, Namirotene, etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Acetylthiophene is used as an intermediate in the manufacturing of drugs like Tiamonium Iodide, Suprofan, Stepronin, Tenonitrozole, Namirotene, etc.
Solubility
Insoluble in water.
Notes
It is sensitive to light. Incompatible with oxidizing agents.
Chemical Identifiers
88-15-3 | |
126.173 | |
WYJOVVXUZNRJQY-UHFFFAOYSA-N | |
6920 | |
CC(=O)C1=CC=CS1 |
C6H6OS | |
MFCD00005442 | |
2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone | |
1-thiophen-2-ylethanone |
Specifications
88-15-3 | |
1.168 | |
91°C (195°F) | |
C6H6OS | |
MFCD00005442 | |
107910 | |
2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone | |
WYJOVVXUZNRJQY-UHFFFAOYSA-N | |
1-thiophen-2-ylethanone | |
6920 | |
99% |
10°C to 12°C | |
213°C to 214°C | |
Slightly Sweetish | |
1.566 | |
UN2810 | |
Light sensitive | |
Insoluble in water. | |
CC(=O)C1=CC=CS1 | |
126.173 | |
126.18 | |
2-Acetylthiophene |
Safety and Handling
GHS H Statement
H300-H311-H330-H227
Fatal if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P370+P378q-P501c
H227-H301+H311+H331
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 201-804-6
missing translation for 'rtecsNumber' : OB6300000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only