missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Allyl-L-glycine, 95%, Thermo Scientific™
£55.50 - £165.00
Chemical Identifiers
CAS | 195316-72-4 |
---|---|
Molecular Formula | C5H10ClNO2 |
Molecular Weight (g/mol) | 151.59 |
MDL Number | MFCD00002627 |
InChI Key | DIDZZOWASZMNQW-WCCKRBBISA-N |
Synonym | s-2-aminopent-4-enoic acid hydrochloride, l-2-allylglycine hydrochloride, s---2-amino-4-pentenoic acid hydrochloride, 2s-2-aminopent-4-enoic acid hydrochloride, 4-pentenoic acid, 2-amino-, hydrochloride, 2s, l-allylglycine hydrochloride, l-alpha-allylglycine hydrochloride, s-2-amino-4-pentenoic acid hydrochloride, 4-pentenoic acid, 2-amino-, hydrochloride, s |
PubChem CID | 44630067 |
IUPAC Name | (2S)-2-aminopent-4-enoic acid;hydrochloride |
SMILES | C=CCC(C(=O)O)N.Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
195316-72-4 | |
151.59 | |
DIDZZOWASZMNQW-WCCKRBBISA-N | |
44630067 | |
C=CCC(C(=O)O)N.Cl |
C5H10ClNO2 | |
MFCD00002627 | |
s-2-aminopent-4-enoic acid hydrochloride, l-2-allylglycine hydrochloride, s---2-amino-4-pentenoic acid hydrochloride, 2s-2-aminopent-4-enoic acid hydrochloride, 4-pentenoic acid, 2-amino-, hydrochloride, 2s, l-allylglycine hydrochloride, l-alpha-allylglycine hydrochloride, s-2-amino-4-pentenoic acid hydrochloride, 4-pentenoic acid, 2-amino-, hydrochloride, s | |
(2S)-2-aminopent-4-enoic acid;hydrochloride |
Specifications
195316-72-4 | |
MFCD00002627 | |
DIDZZOWASZMNQW-WCCKRBBISA-N | |
(2S)-2-aminopent-4-enoic acid;hydrochloride | |
44630067 | |
95% |
C5H10ClNO2 | |
s-2-aminopent-4-enoic acid hydrochloride, l-2-allylglycine hydrochloride, s---2-amino-4-pentenoic acid hydrochloride, 2s-2-aminopent-4-enoic acid hydrochloride, 4-pentenoic acid, 2-amino-, hydrochloride, 2s, l-allylglycine hydrochloride, l-alpha-allylglycine hydrochloride, s-2-amino-4-pentenoic acid hydrochloride, 4-pentenoic acid, 2-amino-, hydrochloride, s | |
C=CCC(C(=O)O)N.Cl | |
151.59 | |
115.13 | |
2-Allyl-L-glycine |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures