missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Aminoheptane, 98+%, Thermo Scientific Chemicals

£31.70 - £81.50

Chemical Identifiers

CAS 123-82-0
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00008101
InChI Key VSRBKQFNFZQRBM-UHFFFAOYNA-N
Synonym 2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil
PubChem CID 5603
IUPAC Name heptan-2-amine
SMILES CCCCCC(C)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11459907
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B22863.14
25 g
£31.70
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11469907
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B22863.22
100 g
£81.50
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

(±)-2-Aminoheptane is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-2-Aminoheptane is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water ( 9 g/L at 20°C).

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
Specifications

Chemical Identifiers

123-82-0
115.22
VSRBKQFNFZQRBM-UHFFFAOYNA-N
5603
CCCCCC(C)N
C7H17N
MFCD00008101
2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil
heptan-2-amine

Specifications

123-82-0
11.45
33°C (91°F)
C7H17N
MFCD00008101
635676
14,9802
Soluble in water ( 9g/L at 20°C).
CCCCCC(C)N
115.22
115.22
(+/-)-2-Aminoheptane
0.768
142°C to 144°C
Amine-like
1.419
UN2733
Air Sensitive
2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil
VSRBKQFNFZQRBM-UHFFFAOYNA-N
heptan-2-amine
5603
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H302+H312+H332-H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 204-655-5

missing translation for 'rtecsNumber' : MQ5425000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only