missing translation for 'onlineSavingsMsg'
Learn More

2-(BOC-Amino)ethanethiol, 97%, Thermo Scientific Chemicals

£56.08 - £177.89

Chemical Identifiers

CAS 67385-09-5
Molecular Formula C7H15NO2S
Molecular Weight (g/mol) 177.26
MDL Number MFCD00274335
InChI Key GSJJCZSHYJNRPN-UHFFFAOYSA-N
Synonym 2-boc-amino ethanethiol, boc-cysteamine, tert-butyl n-2-mercaptoethyl carbamate, tert-butyl n-2-sulfanylethyl carbamate, 2-bocamino ethanethiol, tert-butyl 2-mercaptoethyl carbamate, carbamic acid, 2-mercaptoethyl-, 1,1-dimethylethyl ester, n-boc-aet, tert-butoxy-n-2-sulfanylethyl carboxamide, n-boc-cysteamine
PubChem CID 3017761
IUPAC Name tert-butyl N-(2-sulfanylethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCS
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10668224
View Documents
Thermo Scientific Acros
381080050
5 mL Glass Bottle
£56.08
5mL
Please to purchase this item. Need a web account? Register with us today!
10721914
View Documents
Thermo Scientific Acros
381080250
25 mL Glass Bottle
£177.89
25mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67385-09-5
177.26
GSJJCZSHYJNRPN-UHFFFAOYSA-N
3017761
CC(C)(C)OC(=O)NCCS
C7H15NO2S
MFCD00274335
2-boc-amino ethanethiol, boc-cysteamine, tert-butyl n-2-mercaptoethyl carbamate, tert-butyl n-2-sulfanylethyl carbamate, 2-bocamino ethanethiol, tert-butyl 2-mercaptoethyl carbamate, carbamic acid, 2-mercaptoethyl-, 1,1-dimethylethyl ester, n-boc-aet, tert-butoxy-n-2-sulfanylethyl carboxamide, n-boc-cysteamine
tert-butyl N-(2-sulfanylethyl)carbamate

Specifications

67385-09-5
96.0
Colorless to Yellow
96% min. (GC)
(CH3)3COC(O)NH(CH2)2SH
MFCD00274335
GSJJCZSHYJNRPN-UHFFFAOYSA-N
CC(C)(C)OC(=O)NCCS
177.26
3017761
97%
Low Melting Solid or Viscous Liquid
68.0°C (0.3 mmHg)
100.0
Authentic
C7H15NO2S
1.4730 to 1.4770
2-boc-amino ethanethiol, boc-cysteamine, tert-butyl n-2-mercaptoethyl carbamate, tert-butyl n-2-sulfanylethyl carbamate, 2-bocamino ethanethiol, tert-butyl 2-mercaptoethyl carbamate, carbamic acid, 2-mercaptoethyl-, 1,1-dimethylethyl ester, n-boc-aet, tert-butoxy-n-2-sulfanylethyl carboxamide, n-boc-cysteamine
1.0490g/mL
tert-butyl N-(2-sulfanylethyl)carbamate
109°C
177.27
1.049
2-(BOC-Amino)ethanethiol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • 2-(BOC-Amino)ethanethiol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only