missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-2-methylpropane, 96%, stabilized, Thermo Scientific Chemicals
£30.10 - £535.00
Chemical Identifiers
CAS | 507-19-7 |
---|---|
Molecular Formula | C4H9Br |
Molecular Weight (g/mol) | 137.02 |
MDL Number | MFCD00000125 |
InChI Key | RKSOPLXZQNSWAS-UHFFFAOYSA-N |
Synonym | tert-butyl bromide, t-butyl bromide, trimethylbromomethane, propane, 2-bromo-2-methyl, bromotrimethylmethane, 2-bromoisobutane, tertiarybutyl bromide, tert-butylbromide, 2-methyl-2-bromopropane, 1-bromo-1,1-dimethylethane |
PubChem CID | 10485 |
IUPAC Name | 2-bromo-2-methylpropane |
SMILES | CC(C)(C)Br |
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
10496810
|
Thermo Scientific Acros
159261000 |
100 mL | Glass bottle |
£30.10
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10113020
|
Thermo Scientific Acros
159265000 |
500 mL | Glass bottle |
£119.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11954491
|
Thermo Scientific Acros
159260025 |
2.5 L | Glass bottle |
£535.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
507-19-7 | |
137.02 | |
RKSOPLXZQNSWAS-UHFFFAOYSA-N | |
10485 | |
CC(C)(C)Br |
C4H9Br | |
MFCD00000125 | |
tert-butyl bromide, t-butyl bromide, trimethylbromomethane, propane, 2-bromo-2-methyl, bromotrimethylmethane, 2-bromoisobutane, tertiarybutyl bromide, tert-butylbromide, 2-methyl-2-bromopropane, 1-bromo-1,1-dimethylethane | |
2-bromo-2-methylpropane |
Spezifikation
507-19-7 | |
1.1800g/mL | |
18°C | |
95% min. (GC) | |
(CH3)3CBr | |
MFCD00000125 | |
15,1557 | |
tert-butyl bromide, t-butyl bromide, trimethylbromomethane, propane, 2-bromo-2-methyl, bromotrimethylmethane, 2-bromoisobutane, tertiarybutyl bromide, tert-butylbromide, 2-methyl-2-bromopropane, 1-bromo-1,1-dimethylethane | |
RKSOPLXZQNSWAS-UHFFFAOYSA-N | |
2-bromo-2-methylpropane | |
137.02 | |
137.02 | |
Liquid |
-20.0°C | |
72.0°C to 74.0°C | |
Authentic | |
C4H9Br | |
1.4269 to 1.4289 | |
01,127 | |
1.18 | |
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents | |
CC(C)(C)Br | |
Potassium carbonate | |
10485 | |
96% | |
2-Bromo-2-methylpropane |
Sicherheit und Handhabung
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 208-065-9
Produktidentifikator
Signalwort
Gefahrenkategorie
Gefahrenhinweis
Sicherheitshinweis
- 2-Bromo-2-methylpropane
Signalwort
- Danger
Gefahrenkategorie
- Specific target organ toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Gefahrenhinweis
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- H225-Highly flammable liquid and vapour.
Sicherheitshinweis
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only