missing translation for 'onlineSavingsMsg'
Learn More
2-Butanone, +99%, for spectroscopy, Thermo Scientific Chemicals Product Code.: 10666812

2-Butanone, +99%, for spectroscopy, Thermo Scientific Chemicals

Product Code. 10666812
500 mL, Glass Bottle
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10666812

Brand: Thermo Scientific Acros 213015000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketonShow MoreShow Less
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point -87.0°C
Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 at 350nm, 0.1 at 340nm, 1 at 330nm
CAS Min % 99.0
CAS Max % 100.0
Density 0.8060g/mL
Boiling Point 80.0°C
Quantity 500 mL
Flash Point −7°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3780 to 1.3800
Assay Percent Range 99+%
Residue after Evaporation 0.001% max.
Specific Gravity 0.806
Linear Formula C2H5COCH3
Viscosity 0.42 mPa.s (15°C)
Beilstein 01, IV, 3243
Water 0.1% max.
Fieser 01,678
Merck Index 15, 6143
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils
Formula Weight 72.11
Percent Purity ≥99%
Grade Spectroscopy
Chemical Name or Material 2-Butanone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468288 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number213015000,213010025
Lot NumberA0447020 Certificate TypeCertificate of Analysis Date25/04/2024 Catalog Number213015000,213010025
Lot NumberA0402101 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number213015000,213010025
Lot NumberA0380823 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number213015000,213010025
Lot NumberA0434976 Certificate TypeCertificate of Analysis Date05/10/2021 Catalog Number213015000,213010025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Butanone, +99%, for spectroscopy, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.