Learn More
(2-Carboxyethyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals
£28.00 - £95.00
Chemical Identifiers
CAS | 51114-94-4 |
---|---|
Molecular Formula | C21H20BrO2P |
Molecular Weight (g/mol) | 415.27 |
MDL Number | MFCD00031698 |
InChI Key | BVKRDNIULHRLCO-UHFFFAOYSA-N |
Synonym | 2-carboxyethyl triphenylphosphonium bromide, 2-carboxyethyl triphenylphosponium bromide, phosphonium, 2-carboxyethyl triphenyl-, bromide, 2-carboxyethyl triphenylphosphanium bromide, c21h20o2p.br, 2-carboxyethyl-triphenylphosphonium bromide, 2-carboxyethyl triphenyl phosphanium bromide, 2-carboxyethyl triphenyl phosphonium bromide |
PubChem CID | 2733850 |
IUPAC Name | 2-carboxyethyl(triphenyl)phosphanium;bromide |
SMILES | [Br-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10406832
|
Thermo Scientific Acros
366590050 |
5 g | Glass bottle |
£28.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10324792
|
Thermo Scientific Acros
366590250 |
25 g | Glass bottle |
£95.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)Chemical Identifiers
51114-94-4 | |
415.27 | |
BVKRDNIULHRLCO-UHFFFAOYSA-N | |
2733850 | |
[Br-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C21H20BrO2P | |
MFCD00031698 | |
2-carboxyethyl triphenylphosphonium bromide, 2-carboxyethyl triphenylphosponium bromide, phosphonium, 2-carboxyethyl triphenyl-, bromide, 2-carboxyethyl triphenylphosphanium bromide, c21h20o2p.br, 2-carboxyethyl-triphenylphosphonium bromide, 2-carboxyethyl triphenyl phosphanium bromide, 2-carboxyethyl triphenyl phosphonium bromide | |
2-carboxyethyl(triphenyl)phosphanium;bromide |
Specifications
51114-94-4 | |
100.0 | |
Authentic | |
C21H20BrO2P | |
2-carboxyethyl triphenylphosphonium bromide, 2-carboxyethyl triphenylphosponium bromide, phosphonium, 2-carboxyethyl triphenyl-, bromide, 2-carboxyethyl triphenylphosphanium bromide, c21h20o2p.br, 2-carboxyethyl-triphenylphosphonium bromide, 2-carboxyethyl triphenyl phosphanium bromide, 2-carboxyethyl triphenyl phosphonium bromide | |
[Br-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
415.27 | |
415.26 | |
Chunks and/or Powder |
96.0 | |
White to Yellow | |
97% | |
MFCD00031698 | |
BVKRDNIULHRLCO-UHFFFAOYSA-N | |
2-carboxyethyl(triphenyl)phosphanium;bromide | |
2733850 | |
97% | |
(2-Carboxyethyl)triphenylphosphonium bromide, 97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only