missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyrimidine, 98%, Thermo Scientific Chemicals

£22.40 - £108.00

Chemical Identifiers

CAS 1722-12-9
Molecular Formula C4H3ClN2
Molecular Weight (g/mol) 114.53
MDL Number MFCD00006060
InChI Key UNCQVRBWJWWJBF-UHFFFAOYSA-N
Synonym pyrimidine, 2-chloro, 2-chloro-pyrimidine, chloropyrimidine, 2-chloro pyrimidine, 2-chloro-1,3-diazine, 2-chlorpyrimidin, 2-chloro-pyrimindine, 2chloropyrimidine, chloro-pyrimidine, 2-chloropyrimidin
PubChem CID 74404
IUPAC Name 2-chloropyrimidine
SMILES ClC1=NC=CC=N1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11309639
View Documents
Thermo Scientific Alfa Aesar
A15394.09
10 g
£22.40
10g
Please to purchase this item. Need a web account? Register with us today!
11498346
View Documents
Thermo Scientific Alfa Aesar
A15394.14
25 g
£37.90
25g
Please to purchase this item. Need a web account? Register with us today!
11408356
View Documents
Thermo Scientific Alfa Aesar
A15394.22
100 g
£108.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Chloropyrimidine is a useful reagent in the preparation of antivirals and other biologically active compounds. It was used in the synthesis of 4?-(1,1?-(5-(2-methoxyphenoxy)-[2,2?-bipyrimidine]-4,6-diyl)bis(1H-pyrazol-3,1-diyl)) dianiline fluorescent dye, biosensor for protein assay, 2-amino-4-heteroarylpyrimidines, cis- and trans-octahydropyrrolo[2,3]pyridine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloropyrimidine is a useful reagent in the preparation of antivirals and other biologically active compounds. It was used in the synthesis of 4′-(1,1′-(5-(2-methoxyphenoxy)-[2,2′-bipyrimidine]-4,6-diyl)bis(1H-pyrazol-3,1-diyl)) dianiline fluorescent dye, biosensor for protein assay, 2-amino-4-heteroarylpyrimidines, cis- and trans-octahydropyrrolo[2,3]pyridine derivatives.

Solubility
Slightly soluble in water. 33.3 mg soluble in 1 mL of alcohol.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents. Store in refrigerator.
Specifications

Chemical Identifiers

1722-12-9
114.53
UNCQVRBWJWWJBF-UHFFFAOYSA-N
74404
ClC1=NC=CC=N1
C4H3ClN2
MFCD00006060
pyrimidine, 2-chloro, 2-chloro-pyrimidine, chloropyrimidine, 2-chloro pyrimidine, 2-chloro-1,3-diazine, 2-chlorpyrimidin, 2-chloro-pyrimindine, 2chloropyrimidine, chloro-pyrimidine, 2-chloropyrimidin
2-chloropyrimidine

Specifications

1722-12-9
75°C to 76°C (10 mmHg)
C4H3ClN2
UN2811
Hygroscopic
Slightly soluble in water. 33.3 mg soluble in 1 mL of alcohol.
ClC1=NC=CC=N1
114.53
114.54
2-Chloropyrimidine
64°C to 67°C
98°C (208°F)
MFCD00006060
107171
pyrimidine, 2-chloro, 2-chloro-pyrimidine, chloropyrimidine, 2-chloro pyrimidine, 2-chloro-1,3-diazine, 2-chlorpyrimidin, 2-chloro-pyrimindine, 2chloropyrimidine, chloro-pyrimidine, 2-chloropyrimidin
UNCQVRBWJWWJBF-UHFFFAOYSA-N
2-chloropyrimidine
74404
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H319-H317-H335
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 217-020-2

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only