missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro[1,1-biphenyl]-4-carboxylic acid, 95%, Thermo Scientific™
£21.90 - £246.00
Chemical Identifiers
CAS | 365-12-8 |
---|---|
Molecular Formula | C13H8FO2 |
Molecular Weight (g/mol) | 215.20 |
MDL Number | MFCD00278732 |
InChI Key | SLKZDWAZOKIEEU-UHFFFAOYSA-M |
Synonym | 2'-fluorobiphenyl-4-carboxylic acid, 4-2-fluorophenyl benzoic acid, 2'-fluoro-1,1'-biphenyl-4-carboxylic acid, 2'-fluoro 1,1'-biphenyl-4-carboxylic acid, 2'-fluoro-biphenyl-4-carboxylic acid, maybridge3_006504, amtda094, acmc-1cr23, 4-2-fluorophenyl-benzoic acid, 2'-fluoro-4-biphenylcarboxylic acid |
PubChem CID | 715022 |
SMILES | [O-]C(=O)C1=CC=C(C=C1)C1=CC=CC=C1F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10722133
|
Thermo Scientific Maybridge
SEW05200CB |
250 mg |
£21.90
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10551513
|
Thermo Scientific Maybridge
SEW05200DA |
1 g |
£70.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10402304
|
Thermo Scientific Maybridge
SEW05200DE |
5 g |
£246.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
365-12-8 | |
215.20 | |
SLKZDWAZOKIEEU-UHFFFAOYSA-M | |
715022 |
C13H8FO2 | |
MFCD00278732 | |
2'-fluorobiphenyl-4-carboxylic acid, 4-2-fluorophenyl benzoic acid, 2'-fluoro-1,1'-biphenyl-4-carboxylic acid, 2'-fluoro 1,1'-biphenyl-4-carboxylic acid, 2'-fluoro-biphenyl-4-carboxylic acid, maybridge3_006504, amtda094, acmc-1cr23, 4-2-fluorophenyl-benzoic acid, 2'-fluoro-4-biphenylcarboxylic acid | |
[O-]C(=O)C1=CC=C(C=C1)C1=CC=CC=C1F |
Specifications
365-12-8 | |
C13H8FO2 | |
MFCD00278732 | |
SLKZDWAZOKIEEU-UHFFFAOYSA-M | |
2'-fluoro-[1,1'-biphenyl]-4-carboxylate | |
715022 | |
95% |
95% | |
FC6H4C6H4CO2H | |
2'-fluorobiphenyl-4-carboxylic acid, 4-2-fluorophenyl benzoic acid, 2'-fluoro-1,1'-biphenyl-4-carboxylic acid, 2'-fluoro 1,1'-biphenyl-4-carboxylic acid, 2'-fluoro-biphenyl-4-carboxylic acid, maybridge3_006504, amtda094, acmc-1cr23, 4-2-fluorophenyl-benzoic acid, 2'-fluoro-4-biphenylcarboxylic acid | |
[O-]C(=O)C1=CC=C(C=C1)C1=CC=CC=C1F | |
215.20 | |
216.21 | |
2'-Fluoro[1, 1'-biphenyl]-4-carboxylic acid |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Fluoro[1,1-biphenyl]-4-carboxylic acid, 95%, Thermo Scientific™