missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-methoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

£29.55 - £63.32

Chemical Identifiers

CAS 352303-67-4
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.946
MDL Number MFCD02094728
InChI Key JCKZNMSBFBPDPM-UHFFFAOYSA-N
Synonym 2-fluoro-3-methoxyphenyl boronic acid, 2-fluoro-3-methoxybenzeneboronic acid, 2-fluoro-3-methoxyphenyl boranediol, 2-fluoro-3-methoxyphenylboronicacid, boronic acid, 2-fluoro-3-methoxyphenyl, pubchem1848, 2-fluoro-3-methoxy-phenyl boronic acid, acmc-1af04, 2-fluoro-3-methoxy-phenylboronic acid
PubChem CID 4985744
IUPAC Name (2-fluoro-3-methoxyphenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)OC)F)(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11315475
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
L19655.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11325475
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
L19655.06
5 g
£63.32
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Fluoro-3-methoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.