Learn More
2-Fluoro-6-methoxybenzyl bromide, 98%, Thermo Scientific Chemicals
£42.60 - £122.00
Chemical Identifiers
CAS | 500912-17-4 |
---|---|
Molecular Formula | C8H8BrFO |
Molecular Weight (g/mol) | 219.053 |
MDL Number | MFCD08688579 |
InChI Key | PVKFBCBTHHTDEX-UHFFFAOYSA-N |
Synonym | 2-fluoro-6-methoxybenzyl bromide, 2-bromomethyl-1-fluoro-3-methoxybenzene, 2-bromomethyl-3-fluoroanisole, 2-fluoro-6-methoxy-benzyl bromide, 2-bromomethyl-3-fluorophenyl methyl ether, benzene,2-bromomethyl-1-fluoro-3-methoxy, 2-bromomethyl-1-fluoranyl-3-methoxy-benzene |
PubChem CID | 2779322 |
IUPAC Name | 2-(bromomethyl)-1-fluoro-3-methoxybenzene |
SMILES | COC1=C(C(=CC=C1)F)CBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15443407
|
Thermo Scientific Alfa Aesar
H26136.MD |
250 mg |
£42.60
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11342139
|
Thermo Scientific Alfa Aesar
H26136.03 |
1 g |
£122.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
500912-17-4 | |
219.053 | |
PVKFBCBTHHTDEX-UHFFFAOYSA-N | |
2779322 | |
COC1=C(C(=CC=C1)F)CBr |
C8H8BrFO | |
MFCD08688579 | |
2-fluoro-6-methoxybenzyl bromide, 2-bromomethyl-1-fluoro-3-methoxybenzene, 2-bromomethyl-3-fluoroanisole, 2-fluoro-6-methoxy-benzyl bromide, 2-bromomethyl-3-fluorophenyl methyl ether, benzene,2-bromomethyl-1-fluoro-3-methoxy, 2-bromomethyl-1-fluoranyl-3-methoxy-benzene | |
2-(bromomethyl)-1-fluoro-3-methoxybenzene |
Specifications
500912-17-4 | |
C8H8BrFO | |
MFCD08688579 | |
Moisture sensitive | |
PVKFBCBTHHTDEX-UHFFFAOYSA-N | |
2-(bromomethyl)-1-fluoro-3-methoxybenzene | |
2779322 | |
98% |
18°C to 21°C | |
1.557 | |
UN3265 | |
2-fluoro-6-methoxybenzyl bromide, 2-bromomethyl-1-fluoro-3-methoxybenzene, 2-bromomethyl-3-fluoroanisole, 2-fluoro-6-methoxy-benzyl bromide, 2-bromomethyl-3-fluorophenyl methyl ether, benzene,2-bromomethyl-1-fluoro-3-methoxy, 2-bromomethyl-1-fluoranyl-3-methoxy-benzene | |
COC1=C(C(=CC=C1)F)CBr | |
219.053 | |
219.05 | |
2-Fluoro-6-methoxybenzyl bromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only