Learn More
2-Fluoro-6-nitrotoluene, 98%, Thermo Scientific™
£18.70 - £139.00
Chemical Identifiers
CAS | 769-10-8 |
---|---|
Molecular Formula | C7H6FNO2 |
Molecular Weight (g/mol) | 155.13 |
MDL Number | MFCD00007197 |
InChI Key | GXPIVRKDWZKIKZ-UHFFFAOYSA-N |
Synonym | 2-fluoro-6-nitrotoluene, 1-fluor-2-methyl-3-nitrobenzene, 2-nitro-6-fluorotoluene, 6-fluoro-2-nitrotoluene, benzene, 1-fluoro-2-methyl-3-nitro, 3-fluoro-2-methyl-1-nitrobenzene, 1-fluor-2-methyl-3-nitrobenzol, pubchem1594, acmc-1bc1k |
PubChem CID | 69854 |
IUPAC Name | 1-fluoro-2-methyl-3-nitrobenzene |
SMILES | CC1=C(F)C=CC=C1[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11394526
|
Thermo Scientific Alfa Aesar
L20093.06 |
5 g |
£18.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314536
|
Thermo Scientific Alfa Aesar
L20093.22 |
100 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
769-10-8 | |
155.13 | |
GXPIVRKDWZKIKZ-UHFFFAOYSA-N | |
69854 | |
CC1=C(F)C=CC=C1[N+]([O-])=O |
C7H6FNO2 | |
MFCD00007197 | |
2-fluoro-6-nitrotoluene, 1-fluor-2-methyl-3-nitrobenzene, 2-nitro-6-fluorotoluene, 6-fluoro-2-nitrotoluene, benzene, 1-fluoro-2-methyl-3-nitro, 3-fluoro-2-methyl-1-nitrobenzene, 1-fluor-2-methyl-3-nitrobenzol, pubchem1594, acmc-1bc1k | |
1-fluoro-2-methyl-3-nitrobenzene |
Specifications
769-10-8 | |
1.277 | |
88°C (190°F) | |
1.524 | |
UN2810 | |
2-fluoro-6-nitrotoluene, 1-fluor-2-methyl-3-nitrobenzene, 2-nitro-6-fluorotoluene, 6-fluoro-2-nitrotoluene, benzene, 1-fluoro-2-methyl-3-nitro, 3-fluoro-2-methyl-1-nitrobenzene, 1-fluor-2-methyl-3-nitrobenzol, pubchem1594, acmc-1bc1k | |
CC1=C(F)C=CC=C1[N+]([O-])=O | |
155.13 | |
155.13 | |
2-Fluoro-6-nitrotoluene |
6°C to 8°C | |
208°C to 210°C | |
C7H6FNO2 | |
MFCD00007197 | |
2361978 | |
GXPIVRKDWZKIKZ-UHFFFAOYSA-N | |
1-fluoro-2-methyl-3-nitrobenzene | |
69854 | |
98% |
Safety and Handling
GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 212-203-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures